siRNA Details
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136241.1 | NP_001129713.1 | 893 | |||
GenBank mRNA | BC091122.1 | - | 35 |

More Information
Other Species Targeted

Gene Ontology Categories:
