siRNA Details
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139336.1 | NP_647552.1 | 6 | 474 | ||
XM_006244296.2 | XP_006244358.1 | 6 | 532 | |||
XM_008766746.1 | XP_008764968.1 | 7 | 990 | |||
XM_008766747.1 | XP_008764969.1 | 642 | ||||
XM_008766748.1 | XP_008764970.1 | 7 | 975 | |||
XM_008766749.1 | XP_008764971.1 | 6 | 534 | |||
XM_008766750.1 | XP_008764972.1 | 6 | 519 | |||
XM_008766751.1 | XP_008764973.1 | 7 | 990 | |||
XM_008766752.1 | XP_008764974.1 | 6 | 882 | |||
XM_008766753.1 | XP_008764975.1 | 6 | 867 | |||
GenBank mRNA | AF482705.1 | - | 6 | 474 | ||
BC086988.1 | - | 6 | 485 | 7192831 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
