siRNA Details
Gene Symbol: Tax1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004199.1 | NP_001004199.1 | 9 | 1231 | ||
XM_006236480.2 | XP_006236542.1 | 9 | 1274 | |||
XM_006236481.2 | XP_006236543.1 | 9 | 1717 | |||
XM_006236482.2 | XP_006236544.1 | 9 | 1274 | |||
XM_006236483.2 | XP_006236545.1 | 9 | 1274 | |||
GenBank mRNA | AF509819.1 | - | 3 | 407 | ||
AY318959.1 | - | 1147 | ||||
BC081949.1 | - | 9 | 1231 | 7122283 |

More Information
Other Species Targeted

Gene Ontology Categories:
