siRNA Details
Gene Symbol: Tax1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004199.1 | NP_001004199.1 | 18 | 2376 | ||
XM_006236480.2 | XP_006236542.1 | 18 | 2473 | |||
XM_006236481.2 | XP_006236543.1 | 18 | 2916 | |||
XM_006236482.2 | XP_006236544.1 | 18 | 2470 | |||
XM_006236483.2 | XP_006236545.1 | 18 | 2416 | |||
GenBank mRNA | AF509819.1 | - | 12 | 1552 | ||
AY318959.1 | - | 2274 | ||||
BC081949.1 | - | 18 | 2376 | 7122283 | ||
FQ234714.1 | - | 7 | 720 |
