siRNA Details
Gene Symbol: Fmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144737.2 | NP_653338.2 | 6 | 873 | ||
XM_008769604.1 | XP_008767826.1 | 5 | 1137 | |||
XM_008769605.1 | XP_008767827.1 | 6 | 947 | |||
GenBank mRNA | AF458414.1 | - | 6 | 840 | ||
BC070904.1 | - | 6 | 873 | 7100688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
