siRNA Details
Gene Symbol: Fmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144737.2 | NP_653338.2 | 5 | 661 | ||
XM_008769604.1 | XP_008767826.1 | 4 | 925 | |||
XM_008769605.1 | XP_008767827.1 | 5 | 735 | |||
GenBank mRNA | AF458414.1 | - | 5 | 628 | ||
BC070904.1 | - | 5 | 661 | 7100688 | ||
CK365136.1 | - | 5 | 596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
