siRNA Details
Gene Symbol: Gpsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145469.1 | NP_001138941.1 | 3 | 472 | ||
NM_144745.2 | NP_653346.2 | 13 | 1912 | |||
XM_006233670.2 | XP_006233732.1 | 13 | 2016 | |||
XM_008761620.1 | XP_008759842.1 | 5 | 3772 | |||
GenBank mRNA | AF107723.1 | - | 12 | 2036 | ||
AY136742.1 | - | 2 | 273 | |||
BC086535.1 | - | 3 | 472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
