siRNA Details
Gene Symbol: Gpsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144745.2 | NP_653346.2 | 9 | 1232 | ||
XM_006233670.2 | XP_006233732.1 | 9 | 1336 | |||
GenBank mRNA | AF107723.1 | - | 8 | 1356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
