siRNA Details
Gene Symbol: Cant1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144754.2 | NP_653355.1 | 2 | 454 | ||
XM_006247789.2 | XP_006247851.1 | 2 | 976 | |||
XM_006247790.1 | XP_006247852.1 | 2 | 524 | |||
GenBank mRNA | AJ312207.1 | - | 1 | 409 | ||
BC097279.1 | - | 2 | 454 | 7443057 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
