siRNA Details
Gene Symbol: LOC103690008 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257157.2 | XP_006257219.2 | 2789 | |||
XM_008773351.1 | XP_008771573.1 | 2786 | ||||
XM_008773352.1 | XP_008771574.1 | 7 | 2675 | |||
XM_008773353.1 | XP_008771575.1 | 7 | 2672 |
Gene Symbol: Atrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773366.1 | XP_008771588.1 | 10 | 2449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
