siRNA Details
Gene Symbol: LOC103690008 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257157.2 | XP_006257219.2 | 1054 | |||
XM_008773351.1 | XP_008771573.1 | 1051 | ||||
XM_008773352.1 | XP_008771574.1 | 5 | 940 | |||
XM_008773353.1 | XP_008771575.1 | 4, 5 | 937 |
Gene Symbol: Atrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773366.1 | XP_008771588.1 | 8 | 714 |

More Information
Other Species Targeted

Gene Ontology Categories:
