siRNA Details
Gene Symbol: Gtf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113570.1 | NP_001107042.1 | 3 | 476 | ||
GenBank mRNA | AF391798.1 | - | 3 | 237 | ||
CB616240.1 | - | 3 | 484 |

More Information
Other Species Targeted

Gene Ontology Categories:
