siRNA Details
Gene Symbol: Pdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145091.4 | NP_659559.2 | 2 | 454 | ||
XM_006255048.2 | XP_006255110.1 | 2 | 499 | |||
GenBank mRNA | AF062741.1 | - | 2 | 354 | ||
BC072485.1 | - | 454 | 7105072 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
