siRNA Details
Gene Symbol: Aard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145093.1 | NP_659561.1 | 2 | 393 | ||
GenBank mRNA | AY007690.1 | - | 2 | 393 |

More Information
Other Species Targeted

Gene Ontology Categories:
