siRNA Details
Gene Symbol: Tp63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127342.1 | NP_001120814.1 | 10, 11 | 1367 | ||
NM_019221.3 | NP_062094.1 | 12, 13 | 1796 | |||
XM_008768791.1 | XP_008767013.1 | 13, 14 | 1726 | |||
GenBank mRNA | AJ277446.1 | - | 11, 12 | 1598 | ||
AJ277447.1 | - | 10, 11 | 1367 | |||
Y10258.2 | - | 12, 13 | 1796 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001127264, NM_011641, NM_001127259, AB010152, AK137253, and others.
Gene Ontology Categories:
