siRNA Details
Gene Symbol: Tp63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127339.1 | NP_001120811.1 | 3 | 365 | ||
NM_001127341.1 | NP_001120813.1 | 3 | 365 | |||
NM_019221.3 | NP_062094.1 | 3 | 365 | |||
XM_006248500.2 | XP_006248562.1 | 3 | 446 | |||
XM_008768791.1 | XP_008767013.1 | 4 | 295 | |||
GenBank mRNA | AJ277446.1 | - | 2 | 167 | ||
AJ277448.1 | - | 2 | 167 | |||
AJ277449.1 | - | 3 | 218 | |||
AJ277451.1 | - | 3 | 218 | |||
AJ277452.1 | - | 2 | 167 | |||
Y10258.2 | - | 3 | 365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
