siRNA Details
Gene Symbol: Bcas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762466.1 | XP_008760688.1 | 6 | 1039 | ||
GenBank mRNA | AB029897.1 | - | 4 | 547 | ||
AB180269.1 | - | 4 | 547 | |||
AB180270.1 | - | 4 | 547 | |||
AB180271.1 | - | 4 | 547 | |||
BC094306.1 | - | 4 | 535 | |||
CO405877.1 | - | 2 | 133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
