siRNA Details
Gene Symbol: Cby1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145676.1 | NP_663709.1 | 5 | 411 | ||
XM_006241967.2 | XP_006242029.1 | 6 | 528 | |||
GenBank mRNA | AF393211.1 | - | 5 | 411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
