siRNA Details
Gene Symbol: Ptpn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145683.1 | NP_663716.1 | 7, 8 | 819 | ||
XM_006249820.2 | XP_006249882.1 | 7, 8 | 1294 | |||
XM_006249821.2 | XP_006249883.1 | 7, 8 | 949 | |||
GenBank mRNA | U28356.1 | - | 7, 8 | 819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
