siRNA Details
Gene Symbol: Gdf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749600.3 | XP_003749648.1 | 1 | 994 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008109, U08337, BC034546, AB259648.
Gene Ontology Categories:
