siRNA Details
Gene Symbol: Gdf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749600.3 | XP_003749648.1 | 2 | 1601 | ||
GenBank mRNA | AB183000.1 | - | 1 | 509 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gdf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749600.3 | XP_003749648.1 | 2 | 1601 | ||
GenBank mRNA | AB183000.1 | - | 1 | 509 |