siRNA Details
Gene Symbol: Tcf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032397.1 | NP_001027569.1 | 2 | 709 | ||
GenBank mRNA | BC100106.1 | - | 2 | 709 | 7379660 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
