siRNA Details
Gene Symbol: Rapgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761973.1 | XP_008760195.1 | 15 | 1429 | ||
GenBank mRNA | AB031229.1 | - | 15 | 1605 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001204166, NM_019688, NM_001204165, AB021132, AK203721, and others.
Gene Ontology Categories:
