siRNA Details
Gene Symbol: Lgi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145769.3 | NP_665712.1 | 6 | 619 | ||
GenBank mRNA | AJ487517.1 | - | 6 | 619 | ||
BC089222.1 | - | 6 | 726 | 7320899 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
