siRNA Details
Gene Symbol: Pcdhga11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037153.1 | NP_001032230.1 | 1 | 509 | ||
GenBank mRNA | AY574010.1 | - | 1 | 509 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pcdhga11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037153.1 | NP_001032230.1 | 1 | 509 | ||
GenBank mRNA | AY574010.1 | - | 1 | 509 |