siRNA Details
Gene Symbol: Dync1li1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145772.1 | NP_665715.1 | 8 | 1009 | ||
GenBank mRNA | AF181992.1 | - | 8 | 1009 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146229, AK217382, AK159983, AK205150, BC023347, and others.
Gene Ontology Categories:
