siRNA Details
Gene Symbol: Mxd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145773.1 | NP_665716.1 | 4 | 343 | ||
GenBank mRNA | FQ234290.1 | - | 4 | 333 | ||
U45986.1 | - | 4 | 343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
