siRNA Details
Gene Symbol: Tubg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145778.2 | NP_665721.1 | 4 | 471 | ||
GenBank mRNA | AB015946.1 | - | 4 | 376 | ||
BC070957.1 | - | 4 | 471 | 7105643 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tubg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145778.2 | NP_665721.1 | 4 | 471 | ||
GenBank mRNA | AB015946.1 | - | 4 | 376 | ||
BC070957.1 | - | 4 | 471 | 7105643 |