siRNA Details
Gene Symbol: Dnah12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252671.2 | XP_006252733.1 | 23 | 3363 | ||
XM_006252672.2 | XP_006252734.1 | 22 | 3237 | |||
XM_006252673.2 | XP_006252735.1 | 22 | 3204 | |||
XM_006252674.2 | XP_006252736.1 | 23 | 3363 | |||
XM_008771017.1 | XP_008769239.1 | 23 | 5166 | |||
XM_008771018.1 | XP_008769240.1 | 13 | 1978 | |||
XM_008771019.1 | XP_008769241.1 | 23 | 3363 | |||
XM_008771020.1 | XP_008769242.1 | 23 | 3363 |
