siRNA Details
Gene Symbol: Phtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191102.1 | NP_001178031.1 | 5 | 369 | ||
XM_006233061.2 | XP_006233123.1 | 6 | 2541 | |||
XM_006233063.2 | XP_006233125.1 | 6 | 2541 | |||
XM_006233064.2 | XP_006233126.1 | 5 | 2483 | |||
XM_006233065.2 | XP_006233127.1 | 6 | 2541 |
