siRNA Details
Gene Symbol: Frmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271054.1 | NP_001257983.1 | 11 | 1433 | ||
XM_006240151.2 | XP_006240213.1 | 11 | 1483 | |||
XM_006240152.2 | XP_006240214.1 | 12 | 1563 | |||
XM_006240153.2 | XP_006240215.1 | 11 | 1907 |
