siRNA Details
Gene Symbol: LOC257650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147142.1 | NP_671483.1 | 3 | 304 | ||
GenBank mRNA | AF260582.1 | - | 3 | 304 |

More Information
Other Species Targeted

