siRNA Details
Gene Symbol: Art5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013039.1 | NP_001013057.1 | 5 | 1140 | ||
XM_006229843.2 | XP_006229905.1 | 3 | 726 | |||
XM_006229845.2 | XP_006229907.1 | 3 | 728 | |||
XM_006229846.2 | XP_006229908.1 | 4 | 758 | |||
XM_008759692.1 | XP_008757914.1 | 3 | 670 | |||
XM_008759693.1 | XP_008757915.1 | 3 | 726 | |||
GenBank mRNA | BC083915.1 | - | 5 | 1140 | 7134556 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
