siRNA Details
Gene Symbol: Slc16a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147216.1 | NP_671749.1 | 6 | 1534 | ||
GenBank mRNA | AJ496570.1 | - | 6 | 1534 |

More Information
Other Species Targeted

Gene Ontology Categories:
