siRNA Details
Gene Symbol: Erc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170787.2 | NP_740768.1 | 1 | 130 | ||
GenBank mRNA | AF541925.1 | - | 1 | 130 | ||
AY049038.1 | - | 1 | 188 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177814, AK147213, AY356530, BC056760, AK032385, and others.
Gene Ontology Categories:
