siRNA Details
Gene Symbol: Aptx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148889.2 | NP_683687.1 | 5 | 650 | ||
XM_006238026.2 | XP_006238088.1 | 5 | 1006 | |||
XM_006238027.2 | XP_006238089.1 | 4 | 1042 | |||
XM_006238028.2 | XP_006238090.1 | 4 | 889 | |||
XM_006238029.2 | XP_006238091.1 | 3 | 223 | |||
XM_008763607.1 | XP_008761829.1 | 3 | 901 | |||
GenBank mRNA | AF398235.1 | - | 4 | 396 | ||
BC078716.1 | - | 5 | 650 | 7132044 | ||
FQ214915.1 | - | 1 | 23 |
