siRNA Details
Gene Symbol: Ostf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148892.3 | NP_683690.2 | 7 | 526 | ||
XM_008760287.1 | XP_008758509.1 | 7 | 521 | |||
GenBank mRNA | BC062400.1 | - | 7 | 505 | 5621572 | |
FM048871.1 | - | 7 | 526 | |||
FM078919.1 | - | 1 | 304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
