siRNA Details
Gene Symbol: Tomm20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152935.1 | NP_690918.1 | 3 | 321 | ||
GenBank mRNA | BC078715.1 | - | 3 | 297 | 7127457 | |
D63411.1 | - | 3 | 310 | |||
FQ224957.1 | - | 3 | 338 | |||
FQ230650.1 | - | 3 | 337 | |||
U21871.1 | - | 3 | 321 |

More Information
Other Species Targeted

Gene Ontology Categories:
