siRNA Details
Gene Symbol: Bles03 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024233.2 | NP_001019404.2 | 2 | 409 | ||
NM_001243534.1 | NP_001230463.1 | 2 | 423 | |||
GenBank mRNA | BC093387.1 | - | 2 | 268 | 7389419 | |
CK483724.1 | - | 2 | 265 | |||
CK596744.1 | - | 2 | 409 | |||
FQ233132.1 | - | 2 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
