siRNA Details
Gene Symbol: Gjc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085381.1 | NP_001078850.1 | 840 | |||
GenBank mRNA | AF536559.4 | - | 433 | |||
BC166405.1 | - | 725 | 9021864 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gjc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085381.1 | NP_001078850.1 | 840 | |||
GenBank mRNA | AF536559.4 | - | 433 | |||
BC166405.1 | - | 725 | 9021864 |