siRNA Details
Gene Symbol: Pkig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153469.4 | NP_703199.1 | 3 | 863 | ||
XM_006235518.2 | XP_006235580.1 | 3 | 2104 | |||
XM_006235520.2 | XP_006235582.1 | 4 | 955 | |||
XM_006235521.1 | XP_006235583.1 | 4 | 1047 | |||
XM_006235522.1 | XP_006235584.1 | 3 | 978 | |||
XM_008762469.1 | XP_008760691.1 | 3 | 2325 | |||
XM_008762470.1 | XP_008760692.1 | 3 | 894 | |||
GenBank mRNA | BC088108.1 | - | 3 | 863 | 7368431 | |
FQ218619.1 | - | 3 | 959 |

More Information
Other Species Targeted

Gene Ontology Categories:
