siRNA Details
Gene Symbol: Mnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153472.2 | NP_703202.1 | 5 | 547 | ||
GenBank mRNA | AY135567.1 | - | 5 | 502 | ||
BC078712.1 | - | 5 | 547 | 7128354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
