siRNA Details
Gene Symbol: Krt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153476.1 | NP_703206.1 | 1 | 596 | ||
GenBank mRNA | AY128946.1 | - | 1 | 596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
