siRNA Details
Gene Symbol: Slc12a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153625.1 | NP_705889.1 | 7, 8 | 950 | ||
XM_006248422.2 | XP_006248484.1 | 8, 9 | 1130 | |||
XM_006248423.2 | XP_006248485.1 | 8, 9 | 1095 | |||
XM_006248424.2 | XP_006248486.1 | 8, 9 | 1379 | |||
XM_006248425.2 | XP_006248487.1 | 8, 9 | 968 | |||
XM_006248426.2 | XP_006248488.1 | 7, 8 | 1016 | |||
XM_006248429.2 | XP_006248491.1 | 8, 9 | 1130 | |||
XM_008768704.1 | XP_008766926.1 | 8, 9 | 1130 | |||
XM_008768706.1 | XP_008766928.1 | 8, 9 | 1130 | |||
GenBank mRNA | AB024430.1 | - | 7, 8 | 950 | ||
AB024431.1 | - | 7, 8 | 950 |
