siRNA Details
Gene Symbol: Prop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153627.1 | NP_705891.1 | 1, 2 | 115 | ||
XM_008767664.1 | XP_008765886.1 | 1, 2 | 112 | |||
GenBank mRNA | AB037922.1 | - | 1, 2 | 115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
