siRNA Details
Gene Symbol: Slc17a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153725.1 | NP_714947.1 | 12 | 2180 | ||
XM_006241240.2 | XP_006241302.1 | 13 | 2083 | |||
XM_008765221.1 | XP_008763443.1 | 11 | 1873 | |||
GenBank mRNA | AJ491795.1 | - | 12 | 2180 | ||
AY117026.1 | - | 12 | 1574 |
