siRNA Details
Gene Symbol: Eaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172047.1 | NP_742044.1 | 5 | 655 | ||
XM_008768707.1 | XP_008766929.1 | 5 | 542 | |||
XM_008768708.1 | XP_008766930.1 | 3 | 1211 | |||
XM_008768709.1 | XP_008766931.1 | 3 | 941 | |||
GenBank mRNA | AY049022.1 | - | 5 | 655 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001113405, NM_001113401, NM_134111, AY034479, AY049021, and others.
Gene Ontology Categories:
