siRNA Details
Gene Symbol: Sostdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153737.1 | NP_714959.1 | 1 | 184 | ||
GenBank mRNA | AF411056.1 | - | 1 | 184 | ||
BC081710.1 | - | 1 | 158 | 7191839 | ||
FQ213516.1 | - | 1 | 174 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025312, AB059271, AK002240, AK007967, AK007893, and others.
Gene Ontology Categories:
