siRNA Details
Gene Symbol: Prl7b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153738.1 | NP_714960.1 | 6 | 628 | ||
GenBank mRNA | AF525159.1 | - | 6 | 628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
