siRNA Details
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153821.1 | NP_722543.1 | 2 | 693 | ||
XM_006250151.2 | XP_006250213.1 | 2 | 1431 | |||
GenBank mRNA | AY730764.1 | - | 2 | 417 | ||
S82911.1 | - | 2 | 693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
